Gene Transcripts

Gene Symbol
Hsd17b1
Entrez Gene ID
25322
Gene Name
hydroxysteroid (17-beta) dehydrogenase 1
Gene Aliases
17BHD1
Chromosome Location
Chr. 10: 88987558 - 88989758 on Build Rnor_6.0
UniGene ID
Rn.10594
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012851.2 NP_036983.1 6 1393
GenBank mRNA BC086365.1 - 6 1378
X97754.1 - 6 1394

View Full Product Details