Gene Transcripts

Gene Symbol
Arrb2
Entrez Gene ID
25388
Gene Name
arrestin, beta 2
Gene Aliases
BARRES
Chromosome Location
Chr. 10: 57040252 - 57048045 on Build Rnor_6.0
UniGene ID
Rn.32973
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012911.1 NP_037043.1 14 1318
GenBank mRNA BC087578.1 - 14 1190
M91590.1 - 14 1318
U03627.1 - 4 245

View Full Product Details