Gene Transcripts
- Gene Symbol
- Lhcgr
- Entrez Gene ID
- 25477
- Gene Name
- luteinizing hormone/choriogonadotropin receptor
- Gene Aliases
- LH/CG-R, LSH-R, LSHR, Lhr
- Chromosome Location
- Chr. 6: 12493182 - 12554482 on Build Rnor_6.0
- UniGene ID
- Rn.11216
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012978.1 |
NP_037110.1
|
3 |
334 |
|
GenBank mRNA |
M26199.1 |
- |
3 |
334 |
|
View Full Product Details