Gene Transcripts

Gene Symbol
Msx2
Entrez Gene ID
25483
Gene Name
msh homeobox 2
Gene Aliases
Chromosome Location
Chr. 17: 11683862 - 11689527 on Build Rnor_6.0
UniGene ID
Rn.10414
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012982.3 NP_037114.2 2 548
GenBank mRNA BF414483.1 - 2 321

View Full Product Details