Gene Transcripts
- Gene Symbol
- Sycp3
- Entrez Gene ID
- 25561
- Gene Name
- synaptonemal complex protein 3
- Gene Aliases
- RNASCP3
- Chromosome Location
- Chr. 7: 29029887 - 29044344 on Build Rnor_6.0
- UniGene ID
- Rn.34889
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013041.1 |
NP_037173.1
|
2 |
177 |
|
GenBank mRNA |
X75785.1 |
- |
2 |
177 |
|
View Full Product Details