Gene Transcripts

Gene Symbol
Igfbp2
Entrez Gene ID
25662
Gene Name
insulin-like growth factor binding protein 2
Gene Aliases
IGFBP-2, ILGFBPA
Chromosome Location
Chr. 9: 80118029 - 80144804 on Build Rnor_6.0
UniGene ID
Rn.6813
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013122.2 NP_037254.2 2 505
GenBank mRNA BC092570.1 - 2 505
FQ212531.1 - 2 524
J04486.1 - 2 697
M31672.1 - 2 533

View Full Product Details