Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013127.1 | NP_037259.1 | 1 | 191 | |
XM_006251072.2 | XP_006251134.1 | 1 | 472 | ||
GenBank mRNA | D29646.1 | - | 1 | 191 | |
D30795.1 | - | 1 | 186 | ||
FQ212353.1 | - | 1 | 196 |