Gene Transcripts

Gene Symbol
Slc18a1
Entrez Gene ID
25693
Gene Name
solute carrier family 18 (vesicular monoamine transporter), member 1
Gene Aliases
CGAT, VMAT-1
Chromosome Location
Chr. 16: 22362002 - 22395183 on Build Rnor_6.0
UniGene ID
Rn.89261
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013152.2 NP_037284.2 3 410
XM_006252993.2 XP_006253055.1 2 610
XM_008771067.1 XP_008769289.1 2 610
GenBank mRNA M97380.1 - 3 439

View Full Product Details