Gene Transcripts

Gene Symbol
FBXL3
Entrez Gene ID
26224
Gene Name
F-box and leucine rich repeat protein 3
Gene Aliases
FBL3, FBL3A, FBXL3A
Chromosome Location
Chr. 13: 76992597 - 77027196 on Build GRCh38
UniGene ID
Hs.508284
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012158.2 NP_036290.1 5 3221
XM_005266336.1 XP_005266393.1 5 3004
XM_005266337.2 XP_005266394.1 4 2856
GenBank mRNA AF126028.1 - 4 2825
AK025484.1 - 3 2419
AL833187.1 - 5 3169
BC072448.1 - 5 3175 6166244

View Full Product Details