Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017147.2 | NP_058843.1 | 2 | 183 | |
GenBank mRNA | BC059143.1 | - | 2 | 183 | |
BC086533.1 | - | 2 | 168 | 7305422 | |
FQ212136.1 | - | 1 | 56 | ||
FQ212794.1 | - | 2 | 182 | ||
FQ212910.1 | - | 2 | 178 | ||
FQ213701.1 | - | 2 | 178 | ||
FQ213897.1 | - | 2 | 207 | ||
FQ214120.1 | - | 2 | 179 | ||
FQ214438.1 | - | 2 | 178 | ||
FQ220384.1 | - | 2 | 194 | ||
FQ220571.1 | - | 2 | 180 | ||
FQ226405.1 | - | 2 | 178 | ||
FQ227487.1 | - | 2 | 180 | ||
FQ229073.1 | - | 2 | 207 | ||
FQ229766.1 | - | 2 | 182 | ||
FQ229833.1 | - | 2 | 178 | ||
FQ229875.1 | - | 2 | 182 | ||
FQ230001.1 | - | 2 | 182 | ||
FQ230091.1 | - | 2 | 179 | ||
FQ231379.1 | - | 2 | 178 | ||
FQ232577.1 | - | 1 | 58 | ||
FQ233750.1 | - | 2 | 178 | ||
FQ234162.1 | - | 2 | 182 | ||
FQ234303.1 | - | 2 | 178 | ||
X62908.1 | - | 1 | 107 |