Gene Transcripts

Gene Symbol
Gclm
Entrez Gene ID
29739
Gene Name
glutamate cysteine ligase, modifier subunit
Gene Aliases
Glclr
Chromosome Location
Chr. 2: 225827504 - 225847876 on Build Rnor_6.0
UniGene ID
Rn.2460
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017305.2 NP_059001.1 1, 2 256
GenBank mRNA BC078867.1 - 1, 2 256
L22191.1 - 1, 2 194
S65555.1 - 1, 2 194

View Full Product Details