Gene Transcripts
- Gene Symbol
- Bsn
- Entrez Gene ID
- 29138
- Gene Name
- bassoon (presynaptic cytomatrix protein)
- Gene Aliases
- Znf231
- Chromosome Location
- Chr. 8: 116873721 - 116965396 on Build Rnor_6.0
- UniGene ID
- Rn.29999
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019146.2 |
NP_062019.2
|
2 |
648 |
|
GenBank mRNA |
Y16563.1 |
- |
2 |
648 |
|
View Full Product Details