Gene Transcripts

Gene Symbol
Cyp2c11
Entrez Gene ID
29277
Gene Name
cytochrome P450, subfamily 2, polypeptide 11
Gene Aliases
CYP2CII, Cyp2c
Chromosome Location
Chr. 1: 257676172 - 258004428 on Build Rnor_6.0
UniGene ID
Rn.10870
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019184.2 NP_062057.2 1 153
GenBank mRNA BC088146.1 - 1 143
J02657.1 - 1 153
U33173.1 - 1 153

View Full Product Details