Gene Transcripts

Gene Symbol
Colq
Entrez Gene ID
29755
Gene Name
collagen-like tail subunit (single strand of homotrimer) of asymmetric acetylcholinesterase
Gene Aliases
Chromosome Location
Chr. 16: 7626380 - 7681621 on Build Rnor_6.0
UniGene ID
Rn.87303
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019274.2 NP_062147.1 2 186
GenBank mRNA AF007583.1 - 2 186

View Full Product Details