Gene Transcripts
- Gene Symbol
- Npy4r
- Entrez Gene ID
- 29471
- Gene Name
- neuropeptide Y receptor Y4
- Gene Aliases
- Ppyr1
- Chromosome Location
- Chr. 16: 10489450 - 10490706 on Build Rnor_6.0
- UniGene ID
- Rn.10574
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031581.2 |
NP_113769.1
|
1 |
1247 |
|
GenBank mRNA |
U42388.1 |
- |
1 |
1249 |
|
View Full Product Details