Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012613.1 | NP_036745.1 | 1 | 945 | |
XM_006232591.2 | XP_006232653.1 | 2 | 1446 | ||
GenBank mRNA | BC128742.1 | - | 1 | 931 | |
M74535.1 | - | 2 | 894 | ||
X14773.1 | - | 1 | 802 |