Gene Transcripts

Gene Symbol
Bsg
Entrez Gene ID
25246
Gene Name
basigin (Ok blood group)
Gene Aliases
5A11, CE9, EMMPRIN, HT7, Ox47R, basignin
Chromosome Location
Chr. 7: 12875536 - 12882753 on Build Rnor_6.0
UniGene ID
Rn.2269
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109882.1 NP_001103352.1 8 1404
NM_012783.3 NP_036915.1 7 1056
GenBank mRNA AY120888.1 - 8 1323
BC059145.1 - 7 980 6889785
FQ212571.1 - 7 992
FQ212582.1 - 7 1000
FQ213065.1 - 7 1002
FQ213626.1 - 7 993
FQ214193.1 - 7 992
FQ214232.1 - 7 997
FQ219910.1 - 7 976
FQ219990.1 - 7 991
FQ220920.1 - 7 1000
FQ224837.1 - 7 1002
FQ229144.1 - 7 1002
FQ229755.1 - 7 991
FQ230014.1 - 7 1003
FQ232262.1 - 7 992
FQ234420.1 - 7 1002
FQ234809.1 - 7 1000
FQ234829.1 - 7 992
X54640.1 - 1183
X67215.1 - 7 977

View Full Product Details