Gene Transcripts

Gene Symbol
TRAM2
Entrez Gene ID
9697
Gene Name
translocation associated membrane protein 2
Gene Aliases
Chromosome Location
Chr. 6: 52497402 - 52577064 on Build GRCh38
UniGene ID
Hs.520182
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012288.3 NP_036420.1 11 6041
XM_011515005.2 XP_011513307.1 11 5916
GenBank mRNA CF264873.1 - 1 683
D31762.1 - 11 5965

View Full Product Details