Gene Transcripts

Gene Symbol
NUFIP1
Entrez Gene ID
26747
Gene Name
NUFIP1, FMR1 interacting protein 1
Gene Aliases
NUFIP, bA540M5.1
Chromosome Location
Chr. 13: 44939249 - 44989478 on Build GRCh38
UniGene ID
Hs.525006
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012345.2 NP_036477.2 10 3247
GenBank mRNA AF159548.1 - 10 3204

View Full Product Details