Gene Transcripts

Gene Symbol
Ddn
Entrez Gene ID
25113
Gene Name
dendrin
Gene Aliases
Den
Chromosome Location
Chr. 7: 140479707 - 140486088 on Build Rnor_6.0
UniGene ID
Rn.5444
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030993.1 NP_112255.1 2 1527
XM_008765672.1 XP_008763894.1 3 2087
GenBank mRNA X96589.1 - 2 1579
Y09000.1 - 2 1527

View Full Product Details