Gene Transcripts

Gene Symbol
Cyp4f5
Entrez Gene ID
286905
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 5
Gene Aliases
Chromosome Location
Chr. 7: 14559806 - 14574345 on Build Rnor_6.0
UniGene ID
Rn.10171
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173124.1 NP_775147.1 380
XM_006241051.2 XP_006241113.1 3 449
XM_008765157.1 XP_008763379.1 3 451
XM_008765158.1 XP_008763380.1 3 451
GenBank mRNA BC129085.1 - 3 341
U39207.1 - 380

Gene Symbol
Cyp4f37
Entrez Gene ID
691312
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 37
Gene Aliases
Chromosome Location
Chr. 7: 14589520 - 14614869 on Build Rnor_6.0
UniGene ID
Rn.10171
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271352.1 NP_001258281.1 4 700

View Full Product Details