Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173149.2 | NP_775172.1 | 2 | 835 | |
XM_006246163.2 | XP_006246225.1 | 3 | 480 | ||
XM_008767665.1 | XP_008765887.1 | 4 | 532 | ||
XM_008767666.1 | XP_008765888.1 | 3 | 1117 | ||
GenBank mRNA | AF035963.1 | - | 2 | 836 | |
BC061820.1 | - | 3 | 275 | 5599247 |