Gene Transcripts

Gene Symbol
Rhot2
Entrez Gene ID
287156
Gene Name
ras homolog family member T2
Gene Aliases
Miro2
Chromosome Location
Chr. 10: 15205945 - 15211739 on Build Rnor_6.0
UniGene ID
Rn.8882
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181823.1 NP_861544.1 9 833
GenBank mRNA AY303973.1 - 9 833

View Full Product Details