Gene Transcripts

Gene Symbol
Rtn2
Entrez Gene ID
308410
Gene Name
reticulon 2
Gene Aliases
RTN2-B
Chromosome Location
Chr. 1: 80195594 - 80208449 on Build Rnor_6.0
UniGene ID
Rn.3210
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201562.3 NP_963856.1 2, 3 90
GenBank mRNA AY279211.1 - 2, 3 90
BC129079.1 - 2, 3 205 8370402

View Full Product Details