Gene Transcripts

Gene Symbol
SNX11
Entrez Gene ID
29916
Gene Name
sorting nexin 11
Gene Aliases
Chromosome Location
Chr. 17: 48107533 - 48123064 on Build GRCh38
UniGene ID
Hs.15827
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013323.2 NP_037455.2 3 362
NM_152244.1 NP_689450.1 4 449
XM_005257260.3 XP_005257317.1 3 129
XM_005257261.3 XP_005257318.1 3 396
XM_005257262.3 XP_005257319.1 3 190
XM_011524697.2 XP_011522999.1 4 232
GenBank mRNA AF121861.1 - 3 382
AK023932.1 - 4 449
AK091852.1 - 4 347
AK296569.1 - 4 431
AK298551.1 - 3 185
AK316374.1 - 3 182
BC000768.2 - 3 179 2963905
BC103721.1 - 3 168 6162638
BT006723.1 - 2 95

View Full Product Details