Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005161.4 | NP_005152.1 | 1 | 3674 | |
NR_027991.1 | 2 | 1741 | |||
GenBank mRNA | AK074514.1 | - | 3 | 1183 | |
AK075252.1 | - | 1 | 3422 | ||
AK097232.1 | - | 2 | 3061 | ||
BC032688.1 | - | 2 | 1523 | 5241995 | |
X89271.1 | - | 2 | 1601 |