Gene Transcripts

Gene Symbol
PNCK
Entrez Gene ID
139728
Gene Name
pregnancy up-regulated nonubiquitous CaM kinase
Gene Aliases
BSTK3, CaMK1b
Chromosome Location
Chr. X: 153669723 - 153687568 on Build GRCh38
UniGene ID
Hs.436667
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039582.3 NP_001034671.3 6 890
NM_001135740.1 NP_001129212.1 6 524
XM_005274651.2 XP_005274708.1 6 507
XM_006724809.3 XP_006724872.1 6 716
XM_011531107.2 XP_011529409.2 6 595
XM_011531108.2 XP_011529410.1 6 560
XM_011531110.2 XP_011529412.1 6 612
XM_011531111.2 XP_011529413.1 6 530
XM_011531112.2 XP_011529414.1 6 503
XM_017029277.1 XP_016884766.1 6 815
XM_017029278.1 XP_016884767.1 7 719
XM_017029279.1 XP_016884768.1 6 476
XM_017029280.1 XP_016884769.1 6 507
GenBank mRNA AK296067.1 - 5 811
AK296165.1 - 6 745
AK296203.1 - 6 890
AK303746.1 - 6 524
AK309148.1 - 4 1153
AK309159.1 - 2 637
AK316402.1 - 5 1060
BC064422.1 - 5 582 5751849

View Full Product Details