Gene Transcripts

Gene Symbol
PRSS50
Entrez Gene ID
29122
Gene Name
protease, serine 50
Gene Aliases
CT20, TSP50
Chromosome Location
Chr. 3: 46712116 - 46717883 on Build GRCh38
UniGene ID
Hs.120365
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013270.4 NP_037402.1 4 622
GenBank mRNA AF100707.1 - 4 622
BC033016.1 - 4 613 4822455
BC037775.2 - 4 584 5272458

View Full Product Details