Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001101648.1 | NP_001095118.1 | 2 | 266 | |
NM_001300967.1 | NP_001287896.1 | 2 | 266 | ||
NM_013389.2 | NP_037521.2 | 2 | 266 | ||
XM_011515326.2 | XP_011513628.1 | 2 | 312 | ||
XM_011515327.2 | XP_011513629.1 | 2 | 601 | ||
XM_011515328.2 | XP_011513630.1 | 1 | 139 | ||
GenBank mRNA | AF192522.1 | - | 2 | 266 | |
AF192523.1 | - | 2 | 266 | ||
AY437865.1 | - | 2 | 210 | ||
AY515256.1 | - | 2 | 210 | ||
BC117178.1 | - | 2 | 264 | 40125729 | |
BC143756.1 | - | 2 | 264 | 9052270 | |
FJ481111.1 | - | 2 | 210 |