Gene Transcripts

Gene Symbol
WDR37
Entrez Gene ID
22884
Gene Name
WD repeat domain 37
Gene Aliases
Chromosome Location
Chr. 10: 1056836 - 1132297 on Build GRCh38
UniGene ID
Hs.188495
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014023.3 NP_054742.2 14 3293
GenBank mRNA AB023199.1 - 14 3264
AK125050.1 - 1 2535
AL136827.1 - 14 3271

View Full Product Details