Gene Transcripts

Gene Symbol
C6orf15
Entrez Gene ID
29113
Gene Name
chromosome 6 open reading frame 15
Gene Aliases
STG
Chromosome Location
Chr. 6: 31111223 - 31112555 on Build GRCh38
UniGene ID
Hs.272214
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014070.2 NP_054789.2 2 107
GenBank mRNA AB031481.1 - 2 107
AY358438.1 - 2 111

View Full Product Details