Gene Transcripts

Gene Symbol
ADAMTS3
Entrez Gene ID
9508
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 3
Gene Aliases
ADAMTS-4
Chromosome Location
Chr. 4: 72280969 - 72568799 on Build GRCh38
UniGene ID
Hs.590919
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014243.2 NP_055058.2 3 433
XM_011532421.2 XP_011530723.1 3 465
XM_011532422.2 XP_011530724.1 3 964
GenBank mRNA AB002364.1 - 3 385
AF247668.1 - 3 433
AK295131.1 - 3 353
BC130287.1 - 3 430 40146318
BC132735.1 - 3 430 40146757

View Full Product Details