Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282708.1 | NP_001269637.1 | 3, 4 | 310 | |
NM_014285.6 | NP_055100.2 | 3, 4 | 310 | ||
NR_104230.1 | 3, 4 | 310 | |||
XM_005272176.2 | XP_005272233.1 | 3, 4 | 308 | ||
GenBank mRNA | AK001916.1 | - | 3, 4 | 299 | |
AK022460.1 | - | 3, 4 | 296 | ||
AK296605.1 | - | 3, 4 | 299 | ||
AK303163.1 | - | 3, 4 | 290 | ||
BC000747.2 | - | 3, 4 | 280 | 2821702 | |
BU934039.1 | - | 313 |