Gene Transcripts

Gene Symbol
SEC61G
Entrez Gene ID
23480
Gene Name
Sec61 translocon gamma subunit
Gene Aliases
SSS1
Chromosome Location
Chr. 7: 54752247 - 54759246 on Build GRCh38
UniGene ID
Hs.488282
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012456.1 NP_001012474.1 4 358
NM_014302.3 NP_055117.1 4 362
GenBank mRNA AF054184.1 - 4 357
AF086539.1 - 4 318
BC009480.2 - 4 300 3954682
BC051840.1 - 282 6060540
BU932672.1 - 361
CD385140.1 - 4 336

View Full Product Details