Gene Transcripts

Gene Symbol
ADAM29
Entrez Gene ID
11086
Gene Name
ADAM metallopeptidase domain 29
Gene Aliases
CT73, svph1
Chromosome Location
Chr. 4: 174918358 - 174978180 on Build GRCh38
UniGene ID
Hs.126838
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130703.1 NP_001124175.1 4 1773
NM_001130704.1 NP_001124176.1 4 1748
NM_001130705.1 NP_001124177.1 3 1667
NM_001278125.1 NP_001265054.1 6 1915
NM_001278126.1 NP_001265055.1 6 1908
NM_001278127.1 NP_001265056.1 2 1478
NM_014269.4 NP_055084.3 5 1854
XM_011531556.1 XP_011529858.1 6 1997
XM_011531557.1 XP_011529859.1 7 2155
XM_011531559.1 XP_011529861.1 5 1886
XM_011531560.1 XP_011529862.1 5 1913
XM_011531561.1 XP_011529863.1 4 1805
GenBank mRNA AF134708.1 - 2 1562
AF171929.1 - 1 1184
AF171930.1 - 1 1184
AK292410.1 - 4 1703

View Full Product Details