Gene Transcripts

Gene Symbol
OR10A3
Entrez Gene ID
26496
Gene Name
olfactory receptor family 10 subfamily A member 3
Gene Aliases
HSHTPCRX12, HTPCRX12
Chromosome Location
Chr. 11: 7938576 - 7939520 on Build GRCh38
UniGene ID
Hs.381322
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003745.1 NP_001003745.1 1 810
GenBank mRNA BC137031.1 - 1 884
BC137032.1 - 1 884 9021028

View Full Product Details