Gene Transcripts
- Gene Symbol
- DAPP1
- Entrez Gene ID
- 27071
- Gene Name
- dual adaptor of phosphotyrosine and 3-phosphoinositides 1
- Gene Aliases
- BAM32
- Chromosome Location
- Chr. 4: 99816824 - 99872290 on Build GRCh38
- UniGene ID
- Hs.436271
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001306151.1 |
NP_001293080.1
|
3, 4 |
447 |
|
NM_014395.2 |
NP_055210.2
|
3, 4 |
456 |
|
XM_011531840.2 |
XP_011530142.1
|
3, 4 |
450 |
|
XM_011531842.1 |
XP_011530144.1
|
3, 4 |
450 |
|
XM_017008023.1 |
XP_016863512.1
|
3, 4 |
450 |
|
XM_017008024.1 |
XP_016863513.1
|
3, 4 |
450 |
|
GenBank mRNA |
AF161551.1 |
- |
3, 4 |
431 |
|
AF163254.1 |
- |
3, 4 |
437 |
|
AF178987.1 |
- |
3, 4 |
411 |
|
AF186022.1 |
- |
3, 4 |
447 |
|
BC012924.1 |
- |
3, 4 |
381 |
4517170 |
DN995752.1 |
- |
3, 4 |
467 |
|
View Full Product Details