Gene Transcripts

Gene Symbol
EHMT2
Entrez Gene ID
10919
Gene Name
euchromatic histone lysine methyltransferase 2
Gene Aliases
BAT8, C6orf30, G9A, GAT8, KMT1C, NG36
Chromosome Location
Chr. 6: 31879759 - 31897707 on Build GRCh38
UniGene ID
Hs.709218
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289413.1 NP_001276342.1 13, 14 2076
NM_001318833.1 NP_001305762.1 15, 16 2262
NM_006709.4 NP_006700.3 15, 16 2033
NM_025256.6 NP_079532.5 14, 15 1931
XM_005248824.3 XP_005248881.1 14, 15 2174
XM_006714974.1 XP_006715037.1 14, 15 2178
XM_006714975.1 XP_006715038.1 13, 14 2076
XM_006714976.1 XP_006715039.1 15, 16 2016
XM_017010202.1 XP_016865691.1 14, 15 1914
GenBank mRNA AB209433.2 - 14, 15 2095
AJ315532.1 - 15, 16 2003
AK302904.1 - 13, 14 2076
BC002686.1 - 12, 13 1503
BC009351.1 - 15, 16 2000
BC018718.1 - 15, 16 1991 4123580
BC020970.1 - 14, 15 1968
X69838.1 - 11, 12 1423

View Full Product Details