Gene Transcripts

Gene Symbol
SIT1
Entrez Gene ID
27240
Gene Name
signaling threshold regulating transmembrane adaptor 1
Gene Aliases
SIT, SIT-R
Chromosome Location
Chr. 9: 35649298 - 35650950 on Build GRCh38
UniGene ID
Hs.88012
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014450.2 NP_055265.1 5 474
GenBank mRNA AJ010059.1 - 5 464
AK314758.1 - 5 455
BC102029.1 - 5 472 40031396
BC104491.1 - 5 472 40031403
BC107484.1 - 5 472 40031398
BM919490.1 - 5 314
JQ924063.1 - 4 548

View Full Product Details