Gene Transcripts
- Gene Symbol
- SIGIRR
- Entrez Gene ID
- 59307
- Gene Name
- single Ig and TIR domain containing
- Gene Aliases
- IL-1R8, TIR8
- Chromosome Location
- Chr. 11: 405716 - 417454 on Build GRCh38
- UniGene ID
- Hs.501624
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135053.1 |
NP_001128525.1
|
5, 6 |
739 |
|
NM_001135054.1 |
NP_001128526.1
|
5, 6 |
812 |
|
NM_021805.2 |
NP_068577.2
|
5, 6 |
708 |
|
XM_005253044.2 |
XP_005253101.1
|
5, 6 |
812 |
|
XM_005253045.1 |
XP_005253102.1
|
5, 6 |
708 |
|
XM_005253046.1 |
XP_005253103.1
|
5, 6 |
796 |
|
XM_005253048.4 |
XP_005253105.1
|
3, 4 |
502 |
|
XM_017018099.1 |
XP_016873588.1
|
5, 6 |
812 |
|
GenBank mRNA |
AK025099.1 |
- |
5, 6 |
708 |
|
AK172830.1 |
- |
5, 6 |
708 |
|
AY358342.1 |
- |
4, 5 |
585 |
|
BC003591.1 |
- |
4, 5 |
638 |
2821373 |
BC025953.1 |
- |
5, 6 |
667 |
3939162 |
BC106007.1 |
- |
5, 6 |
789 |
6092503 |
CR457338.1 |
- |
3, 4 |
482 |
|
View Full Product Details