Gene Transcripts

Gene Symbol
HTN3
Entrez Gene ID
3347
Gene Name
histatin 3
Gene Aliases
HIS2, HTN2, HTN5
Chromosome Location
Chr. 4: 70028413 - 70036538 on Build GRCh38
UniGene ID
Hs.654442
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000200.2 NP_000191.1 6 327
GenBank mRNA BC009791.1 - 6 285
BC095438.1 - 6 284 30352697
BP332860.1 - 6 281
CB957931.1 - 6 327
M18372.1 - 5 247
M26665.1 - 6 281

View Full Product Details