Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173800.4 | NP_776161.3 | 14 | 2367 | |
GenBank mRNA | AK075131.1 | - | 9 | 902 | |
AK291655.1 | - | 14 | 2381 | ||
AY560010.1 | - | 14 | 2255 | ||
BC036440.1 | - | 9 | 1246 | ||
BC060869.1 | - | 15 | 2294 | 30346049 | |
BC070028.1 | - | 9 | 1364 | ||
BC094716.1 | - | 9 | 1364 | ||
BC109022.1 | - | 14 | 2231 | 40008501 | |
BC109023.1 | - | 14 | 2231 | 40008502 |