Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001014809.2 | NP_001014809.1 | 14 | 2442 | |
NM_001288661.1 | NP_001275590.1 | 14 | 2549 | ||
NM_001288662.1 | NP_001275591.1 | 14 | 2372 | ||
NM_001313.4 | NP_001304.1 | 14 | 2336 | ||
GenBank mRNA | AK091696.1 | - | 2 | 1720 | |
AK094725.1 | - | 14 | 2101 | ||
AK122754.1 | - | 14 | 2372 | ||
AK127754.1 | - | 15 | 2088 | ||
AK309379.1 | - | 791 | |||
BC000252.1 | - | 14 | 2201 | 3356888 | |
BC007613.1 | - | 14 | 2201 | 3354024 | |
BC007898.2 | - | 10 | 1484 | ||
D78012.1 | - | 14 | 2136 | ||
U17278.1 | - | 12 | 1813 |