Gene Transcripts

Gene Symbol
UAP1
Entrez Gene ID
6675
Gene Name
UDP-N-acetylglucosamine pyrophosphorylase 1
Gene Aliases
AGX, AGX1, AGX2, SPAG2
Chromosome Location
Chr. 1: 162561440 - 162599843 on Build GRCh38
UniGene ID
Hs.492859
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324113.1 NP_001311042.1 6 1015
NM_001324114.1 NP_001311043.1 5 903
NM_001324115.1 NP_001311044.1 6 1242
NM_001324116.1 NP_001311045.1 5 1130
NM_001324117.1 NP_001311046.1 5 1130
NM_003115.4 NP_003106.3 5 1064
XM_011509913.2 XP_011508215.1 6 963
XM_011509914.2 XP_011508216.1 6 1015
GenBank mRNA AB011004.1 - 4 735
AK312685.1 - 5 990
BC009377.2 - 5 1037 4121439
S73498.1 - 5 967

View Full Product Details