Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014470.3 | NP_055285.1 | 4 | 434 | |
GenBank mRNA | AB040147.1 | - | 4 | 412 | |
AF498967.1 | - | 4 | 331 | ||
AK292762.1 | - | 4 | 434 | ||
BC026356.1 | - | 4 | 434 | 4793442 | |
Y07923.1 | - | 4 | 331 |