Gene Transcripts

Gene Symbol
OR52E2
Entrez Gene ID
119678
Gene Name
olfactory receptor family 52 subfamily E member 2
Gene Aliases
Chromosome Location
Chr. 11: 5058650 - 5059627 on Build GRCh38
UniGene ID
Hs.553557
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005164.2 NP_001005164.2 1 424

View Full Product Details