Gene Transcripts

Gene Symbol
OR8D2
Entrez Gene ID
283160
Gene Name
olfactory receptor family 8 subfamily D member 2 (gene/pseudogene)
Gene Aliases
JCG2
Chromosome Location
Chr. 11: 124319262 - 124320197 on Build GRCh38
UniGene ID
Hs.504212
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002918.1 NP_001002918.1 1 618
GenBank mRNA AF162668.1 - 1 618
BC137043.1 - 1 709 9021039
BC137045.1 - 1 709 9021041

View Full Product Details