Gene Transcripts

Gene Symbol
DPH3
Entrez Gene ID
285381
Gene Name
diphthamide biosynthesis 3
Gene Aliases
DELGIP, DELGIP1, DESR1, DPH3A, KTI11, ZCSL2
Chromosome Location
Chr. 3: 16257061 - 16264989 on Build GRCh38
UniGene ID
Hs.388087
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206831.2 NP_996662.1 2 261
GenBank mRNA AK092040.1 - 2 214
AK311618.1 - 2 223
BC010181.2 - 2 170 4543414
DA875039.1 - 2 261

View Full Product Details