Gene Transcripts

Gene Symbol
OR8D4
Entrez Gene ID
338662
Gene Name
olfactory receptor family 8 subfamily D member 4
Gene Aliases
OR11-275
Chromosome Location
Chr. 11: 123906432 - 123907376 on Build GRCh38
UniGene ID
Hs.449688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005197.1 NP_001005197.1 1 139
GenBank mRNA BC137180.1 - 1 180
BC137206.1 - 1 180 9021203

View Full Product Details