Gene Transcripts

Gene Symbol
OR2A12
Entrez Gene ID
346525
Gene Name
olfactory receptor family 2 subfamily A member 12
Gene Aliases
OR2A12P, OR2A16P
Chromosome Location
Chr. 7: 144095108 - 144096040 on Build GRCh38
UniGene ID
Hs.553708
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004135.1 NP_001004135.1 1 16
GenBank mRNA BC136902.1 - 1 76
BC136903.1 - 1 76 9020894

View Full Product Details